Anti-CD23 Rat Monoclonal Antibody (Brilliant Violet® 510) [clone: B3B4]

Supplier: BioLegend
101623 101623
10759-044EA 283.15 USD
CA10759-044 10759-044
Anti-CD23 Rat Monoclonal Antibody (Brilliant Violet® 510) [clone: B3B4]
Antibodies
Brilliant Violet 510™ anti-mouse CD23 [B3B4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD23
Clonality: Monoclonal
Clone: B3B4
Conjugation: Brilliant Violet® 510
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR