Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD213a2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: SHM38]

Supplier: Biolegend

354405 354406 354405 354406
CA10759-742 10759-744 10759-742 CA10759-744
Anti-CD213a2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: SHM38]
Antibodies
APC anti-human CD213α2 (IL13Rα2) [SHM38]; Isotype: Mouse IgG1, κ; Reactivity: Human, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD213a2
Clonality: Monoclonal
Clone: SHM38
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Rhesus

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty