Anti-CD213a2 Mouse Monoclonal Antibody [clone: SHM38]

Supplier: BioLegend
354402 354402 354401 354401
10759-740EA 319.89 USD
10759-740 CA10759-740 10759-738 CA10759-738
Anti-CD213a2 Mouse Monoclonal Antibody [clone: SHM38]
Antibodies
Purified anti-human CD213α2 (IL13Rα2) [SHM38]; Isotype: Mouse IgG1, κ; Reactivity: Human, Rhesus; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD213a2
Clonality: Monoclonal
Clone: SHM38
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR