Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD209B Rat Monoclonal Antibody [clone: 2C7B27]

Supplier: Biolegend

147802 147801 147802 147801
CA10756-628 10756-626 10756-628 CA10756-626
Anti-CD209B Rat Monoclonal Antibody [clone: 2C7B27]
Purified anti-mouse CD209B [2C7B27]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: WB; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.5 - 2.5 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD209B
Clonality: Monoclonal
Clone: 2C7B27
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty