Anti-CD207 Mouse Monoclonal Antibody [clone: 4C7]

Supplier: BioLegend
144201 144202 144202 144201
10759-332EA 109.54 USD
10759-332 CA10759-332 10759-334 CA10759-334
Anti-CD207 Mouse Monoclonal Antibody [clone: 4C7]
Antibodies
Purified anti-mouse/human CD207 (Langerin) [4C7]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse, Human, Cross-Reactivity: Cynomolgus, Rabbit, Swine (Pig, Porcine); Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD207
Clonality: Monoclonal
Clone: 4C7
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Mouse;Human;Cross-Reactivity: Cynomolgus;Rabbit;Swine (Pig;Porcine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR