Anti-CD206 Rat Monoclonal Antibody (Brilliant Violet® 605) [clone: C068C2]

Supplier: BioLegend
141721 141721
10757-254EA 311.31 USD
CA10757-254 10757-254
Anti-CD206 Rat Monoclonal Antibody (Brilliant Violet® 605) [clone: C068C2]
Antibodies
Brilliant Violet 605™ anti-mouse CD206 (MMR) [C068C2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC, FC; Size: 125 μl

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD206
Clonality: Monoclonal
Clone: C068C2
Conjugation: Brilliant Violet® 605
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR