Anti-LYPD6B Mouse Monoclonal Antibody [clone: 634CT7.10.1]

Supplier: Abcepta
AM2160A
89511-954EA 637.11 USD
89511-954
Anti-LYPD6B Mouse Monoclonal Antibody [clone: 634CT7.10.1]
Antibodies
Western Blot: 1:500~4000,

Type: Primary
Antigen: LYPD6B
Clonality: Monoclonal
Clone: 634CT7.10.1
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR