Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD206 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: C068C2]

Supplier: Biolegend

141717 141717
CA10757-248 10757-248
Anti-CD206 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: C068C2]
Brilliant Violet 421™ anti-mouse CD206 (MMR) [C068C2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC, FC, IHC, IF; Size: 125 μl

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. For immunohistochemistry, a concentration range of 2.5-5 μg/ml is suggested. For immunofluorescence microscopy, a concentration range of 2.5-10 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD206
Clonality: Monoclonal
Clone: C068C2
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty