Anti-CD206 Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: C068C2]

Supplier: BioLegend
141715 141716 141716 141715
10757-246EA 424.06 USD
10757-246 CA10757-246 CA10757-244 10757-244
Anti-CD206 Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: C068C2]
Antibodies
PerCP/Cy5.5 anti-mouse CD206 (MMR) [C068C2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC, FC; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD206
Clonality: Monoclonal
Clone: C068C2
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR