Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD205 Mouse Monoclonal Antibody [clone: HD83]

Supplier: Biolegend

359202 359202
10760-558EA 267.72 USD InStock
10760-558 CA10760-558
Anti-CD205 Mouse Monoclonal Antibody [clone: HD83]
Antibodies
Purified anti-human/mouse/rat CD205 (DEC-205) [HD83]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat, Cross Reactivity: Monkey; Apps: FC, IHC, WB; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD205
Clonality: Monoclonal
Clone: HD83
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Mouse;Rat;Cross Reactivity: Monkey

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty