Anti-CD203c (E-NPP3) Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: NP4D6]

Supplier: BioLegend
324620 324620
10758-324EA 481.83 USD
10758-324 CA10758-324
Anti-CD203c (E-NPP3) Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: NP4D6]
Antibodies
Brilliant Violet 605™ anti-human CD203c (E-NPP3) [NP4D6]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD203c (E-NPP3)
Clonality: Monoclonal
Clone: NP4D6
Conjugation: Brilliant Violet® 605
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR