Anti-CD200 R Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: OX-108]

Supplier: Biolegend

329308 329307 329308 329307
10758-394EA 274.91 USD
10758-394 CA10758-392 CA10758-394 10758-392
Anti-CD200 R Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: OX-108]
Antibodies
APC anti-human CD200 Receptor [OX-108]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD200 R
Clonality: Monoclonal
Clone: OX-108
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Baboon
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR