Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD20 Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: 2H7]

Supplier: Biolegend

302336 302335 302336 302335
10757-718 302335-BL CA10757-718 CA302335-BL
Anti-CD20 Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: 2H7]
Brilliant Violet 650™ anti-human CD20 [2H7]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Baboon, Capuchin Monkey, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus, Squirrel Monkey; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: 2H7
Conjugation: Brilliant Violet® 650
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Baboon;Capuchin Monkey;Chimpanzee;Cynomolgus;Pigtailed Macaque;Rhesus;Squirrel Monkey

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty