Anti-CD2 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: RM2-5]

Supplier: BioLegend
100114 100113 100113 100114
10756-678EA 316.31 USD
CA10756-678 CA10756-676 10756-678 10756-676
Anti-CD2 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: RM2-5]
Antibodies
PE/Cy7 anti-mouse CD2 [RM2-5]; Isotype: Rat IgG2b, λ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RM2-5
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: Rat IgG2b, λ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR