Anti-CD1d Mouse Monoclonal Antibody [clone: WTH-1]

Supplier: BioLegend
205802 205802
205802-BLEA 139.32 USD
CA205802-BL 205802-BL
Anti-CD1d Mouse Monoclonal Antibody [clone: WTH-1]
Antibodies
Purified anti-rat CD1d [WTH-1]; Isotype: Mouse IgG2a, κ; Reactivity: Rat; Apps: FC, IHC, IP, WB; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.06 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD1d
Clonality: Monoclonal
Clone: WTH-1
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR