Anti-CD1d Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: K253]

Supplier: BioLegend
140805 140805
140805-BLEA 105.76 USD
CA140805-BL 140805-BL
Anti-CD1d Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: K253]
Antibodies
PE anti-mouse CD1d [K253]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD1d
Clonality: Monoclonal
Clone: K253
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR