Anti-CD1d Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 51.1]

Supplier: BioLegend
350310 350309 350310 350309
10758-960EA 412.26 USD
10758-960 CA10758-960 CA10758-958 10758-958
Anti-CD1d Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 51.1]
Antibodies
PE/Cy7 anti-human CD1d [51.1]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cynomolgus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD1d
Clonality: Monoclonal
Clone: 51.1
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR