Anti-OR6Q1 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP14488B
89521-048EA 636.61 USD
89521-048
Anti-OR6Q1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: OR6Q1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR