Anti-CD199 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: L053E8]

Supplier: BioLegend
358908 358907 358908 358907
10760-490EA 377.86 USD
CA10760-488 10760-490 CA10760-490 10760-488
Anti-CD199 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: L053E8]
Antibodies
APC anti-human CD199 (CCR9) [L053E8]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD199
Clonality: Monoclonal
Clone: L053E8
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR