Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD197 (CCR7) Mouse Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: G043H7]

Supplier: Biolegend

353211 353212 353211 353212
CA353211-BL CA353212-BL 353211-BL 353212-BL
Anti-CD197 (CCR7) Mouse Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: G043H7]
Antibodies
APC/Cy7 anti-human CD197 (CCR7) [G043H7]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. <strong>Test size products are transitioning from 20 μl to 5 μl per test</strong>. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at <a href="/testsize" target="_blank">www.biolegend.com/testsize</a> regarding the test size change.

Type: Primary
Antigen: CD197 (CCR7)
Clonality: Monoclonal
Clone: G043H7
Conjugation: APC (Allophycocyanin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty