Anti-CD196 (CCR6) Armenian Hamster Monoclonal Antibody (Brilliant Violet® 605) [clone: 29-2L17]

Supplier: BioLegend
129819 129819
10757-152EA 334.7 USD
CA10757-152 10757-152
Anti-CD196 (CCR6) Armenian Hamster Monoclonal Antibody (Brilliant Violet® 605) [clone: 29-2L17]
Antibodies
Brilliant Violet 605™ anti-mouse CD196 (CCR6) [29-2L17]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD196 (CCR6)
Clonality: Monoclonal
Clone: 29-2L17
Conjugation: Brilliant Violet® 605
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR