Anti-OR4K14 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP14989A
89520-646EA 636.79 USD
89520-646
Anti-OR4K14 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: OR4K14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR