Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD195 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: J418F1]

Supplier: Biolegend

359120 359119 359119 359120
10760-544EA 245.86 USD InStock
10760-544 10760-542 CA10760-542 CA10760-544
Anti-CD195 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: J418F1]
Antibodies
FITC anti-human CD195 (CCR5) [J418F1]; Isotype: Rat IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD195
Clonality: Monoclonal
Clone: J418F1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty