Anti-REPS2 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP13131A
89512-890EA 636.79 USD
89512-890
Anti-REPS2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000, Immunhistochemsitry: 1:10~50,

Type: Primary
Antigen: REPS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR