Anti-CD195 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: J418F1]

Supplier: BioLegend
359106 359105 359106 359105
10760-516EA 340.51 USD
CA10760-514 10760-516 CA10760-516 10760-514
Anti-CD195 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: J418F1]
Antibodies
PE anti-human CD195 (CCR5) [J418F1]; Isotype: Rat IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD195
Clonality: Monoclonal
Clone: J418F1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR