Anti-PPHLN1 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP20529A
10217-816EA 580.86 USD
10217-816
Anti-PPHLN1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000, Immunofluorescence: 1:25,

Type: Primary
Antigen: PPHLN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR