Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD194 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: L291H4]

Supplier: Biolegend

359419 359420 359420 359419
10760-602EA 131.13 USD InStock
10760-602 CA10760-604 10760-604 CA10760-602
Anti-CD194 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: L291H4]
Antibodies
PE/Dazzle™ 594 anti-human CD194 (CCR4) [L291H4]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD194
Clonality: Monoclonal
Clone: L291H4
Conjugation: PE (Phycoerythrin)/Dazzle 594
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Cynomolgus

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty