Anti-CD193 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 5E8]

Supplier: BioLegend
310714 310714
10758-000EA 448.24 USD
CA10758-000 10758-000
Anti-CD193 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 5E8]
Antibodies
Brilliant Violet 421™ anti-human CD193 (CCR3) [5E8]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD193
Clonality: Monoclonal
Clone: 5E8
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR