Anti-HIPK4 Mouse Monoclonal Antibody [clone: 292CT1.4.1]

Supplier: Abcepta
AM1939B
89520-998EA 637.11 USD
89520-998
Anti-HIPK4 Mouse Monoclonal Antibody [clone: 292CT1.4.1]
Antibodies
Western Blot: 1:100~500,

Type: Primary
Antigen: HIPK4
Clonality: Monoclonal
Clone: 292CT1.4.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR