Anti-CNNM3 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP19487C
89514-818EA 636.79 USD
89514-818
Anti-CNNM3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: CNNM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR