Anti-CD191 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 5F10B29]

Supplier: BioLegend
362906 362905 362905 362906
10760-922EA 450.89 USD
CA10760-920 CA10760-922 10760-922 10760-920
Anti-CD191 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 5F10B29]
Antibodies
Alexa Fluor® 488 anti-human CD191 (CCR1) [5F10B29]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. * Alexa Fluor® 488 has a maximum emission of 519 nm when it is excited at 488 nm.

Type: Primary
Antigen: CD191
Clonality: Monoclonal
Clone: 5F10B29
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR