Anti-LSM14A Rabbit Polyclonal Antibody

Supplier: Abcepta
AP14733B
89512-506EA 636.79 USD
89512-506
Anti-LSM14A Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000, Immunhistochemsitry: 1:10~50,

Type: Primary
Antigen: LSM14A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR