Anti-CD19 Rat Monoclonal Antibody (Alexa Fluor® 594) [clone: 6D5]

Supplier: BioLegend
115552 115552
10756-926EA 332.3 USD
CA10756-926 10756-926
Anti-CD19 Rat Monoclonal Antibody (Alexa Fluor® 594) [clone: 6D5]
Antibodies
Alexa Fluor® 594 anti-mouse CD19 [6D5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IF, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescence staining. For immunohistochemisty, a concentration range of 1-5 μg/ml is suggested. For immunofluorescence microscopy, a concentration range of 1-10 μg/ml is recommended. <span>For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume.</span> It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: 6D5
Conjugation: Alexa Fluor® 594
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR