Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

General-Purpose Undercounter Refrigerator, Thermo Scientific

Supplier: Thermo Scientific
MR05PA-SEEE-TS
89236-760
General-Purpose Undercounter Refrigerator, Thermo Scientific
This undercounter refrigerator keeps critical samples close to the workplace without the worry of temperature variations.

  • Positive locking door latch
  • Hospital grade plugs
  • Stainless steel interior
  • Digital temperature display
  • Standard 1" (25 mm) access port
  • Designed to meet JCAHO guidelines
  • Temperature range of 2° to 10°C, preset to 4°C

Refrigerator has excellent temperature stability (<3°C ensuring no region of the cabinet can reach zero degrees/freezing), heat rejection and noise levels. Features a non-invasive auto defrost mechanism and low energy consumption. Refrigerator is compatible with Pyxis adapters and remote wireless monitoring solutions.

Certifications: UL certified

Order Now
 

ORDER

SERVICES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty