DuPont™ Tyvek® IsoClean® Hood and Mask
Supplier: DuPont
Tyvek®
9820
CV9820CS 1642.11 USD
CV9820
DuPont™ Tyvek® IsoClean® Hood and Mask
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • Ties with loops for adjustable fit
  • One size fits most
  • White hood and blue face mask
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration. Mask is 7" size with pleated polyethylene outer. Garments are gamma sterilized to SAL of 10-6.
Packaging: Individually packaged in an opaque bag.