Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Professional Touch Stirrer with NIST Traceable Certificate, 120V, Talboys

Supplier: TALBOYS ENGINEERING/TROEMNER
984TA7CSTTSUSC
  • Intuitive 4.3” color touch screen
  • Programmable for 5 separate, 5 step programs
  • Visibly track program progress through the live status bar
  • On-board help screens
  • Programmable speed ramping feature (4 options)
  • Includes NIST traceable certificate for speed and time
  • 5 year warranty

The Talboys Professional Touch Stirrers are designed for applications that require exceptional accuracy, stability, and repeatability. Exclusive safety features help protect operator and sample. Stirring function, with continuous duty motor and powerful magnet, maintains set speed even under changing load or viscosity.

Advanced microprocessor regulates the stirring function. Precise speed control provides consistent stirring at all speeds. Programmable speed ramping feature allows user greater control over their assay when changing parameters within a program. Powerful continuous duty motor delivers dependable and reliable stirring. LCD touch screen enables faster setting of speed and time, which can be viewed at once. Display features on-screen help topics. Touch screen is compatible with rubber gloves used in labs. Program control capabilities allow user programmable operation for automated use and memory. Storage for five separate 5-step programs, or unlimited number of programs with the use of the USB. USB port can transfer data to a flash drive for data logging, program storage, and software updates. LCD touch screen enables faster setting of temperature, speed, and time, which can be viewed at once. Display features on-screen help topics. Touch screen is compatible with rubber gloves used in labs.

Low profile design takes up minimal bench space and fits easily into fume hoods. Rear housing features an off centered, built-in support rod holder with locking knob. Top plate is ceramic and features a 7 x 7” (17.8 x 17.8cm) chemical resistant, reflective surface that is easy to clean.

Stirrers are equipped with a variety of innovative safety features designed to facilitate efficient operation and workplace safety. The unit’s housing is constructed of a heat and chemical resistant polymer that remains cool to the touch throughout use. The spill-resistant housing channels fluids away from internal components in the event of a spill. In timed mode, an alarm will sound when time reaches zero. The unit is equipped with a pulse mode feature ideal for quick stirring. Speed ramping feature slowly increases motor speed for improved safety and enhanced coupling. Avoids splashing, improves spin bar control, and provides excellent low end speed control.

Certifications: TUV listed. RoHS compliant.

Ordering information: Units include a 92” (234cm) detachable, 3-wire cord and plug. Supplied with a 1.5” (3.8cm) PTFE coated stir bar. 5 year limited warranty on parts and labor. NIST traceable calibration certificate by an ISO/IEC 17025 accredited laboratory. Multiple data points within the speed and time function are reported on the certificate along with the associated uncertainties.



 

ORDER

Expand All / Collapse All
Electrical Maximum Capacity Speed Range Supplier No. VWR Catalog Number Unit Price Quantity
120V, 50W, 0.4A 2500 mL 60 to 1600 rpm 984TA7CSTTSUSC 10822-488 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Speed stability +/-2%
Timer 1 min to 99 hr. 59 min.
Shipping Weight 5.4 kg (12 lbs.)
Overall Dimensions 14.8L x 9.9W x 4.3H"
Top plate dimensions 17.8 x 12.8 cm (7 x 7")
Top plate material Ceramic

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty