Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Touch Microplate Shakers and Incubating Microplate Shakers with NIST Certificate, 230V, Talboys

Supplier: TALBOYS ENGINEERING/TROEMNER
980TAIMPTSEUC 980TAIMOTSEUC 980TAOMPTSEUC
The Talboys Advanced and Professional Touch 1000-MP Open Air and Incubating Orbital Shakers are designed for a wide variety of heating and shaking applications that require consistent and precise results

  • Incubating or open air models
  • Intuitive 4.3” color touch screen
  • Programmable for 5 separate, 5 step programs
  • Visibly track program progress through the live status bar
  • On-board help screens
  • Holds 4 microplates, deep well plates, or 2 microtube racks
  • Includes NIST traceable certificate for temperature (incubating model), speed, and time
  • 5 year warranty

The variable speed microprocessor control provides consistent uniform shaking action. Microprocessor will display last set-point and will restart if power is interrupted (continuous mode). LCD touch screen enables faster setting of temperature (incubating model), speed and time which can be viewed at once. Display features on-screen help topics. Touch screen is compatible with rubber gloves used in labs. Program control capabilities allow user programmable operation for automated use and memory. Storage for five separate 5-step programs, or unlimited number of programs with the use of the USB. USB port can transfer data to a flash drive for data logging, program storage, and software updates. The unit is equipped with a pulse mode feature ideal for quick mixing. Speed ramping feature ensures that shaker slowly increases speed to desired set-point to avoid splashing.

The triple eccentric drive features permanently lubricated ball bearings. The maintenance free, brushless DC motor provide reliable service and continuous duty operation. Audible and visual signals will activate when system detects an obstruction or overload on the tray. Audible alarm will sound in timed mode when time reaches zero. Spill resistant design channels fluids away from internal components.

(Incubating model) PID temperature controller maintains precise temperature control from ambient +5°C to 65°C. Easy to use temperature adjustments in 1°C increments. Temperature calibration mode allows user to calibrate unit to an external temperature device for up to 6 separate points. Maximum temperature limiting feature ensures the temperature will not exceed user defined limits, allowing the user control of temperature sensitive samples. Caution hot indicator Symbol illuminates when the temperature of the air in the chamber reaches 40°C and remains lit until the temperature cools down. Polycarbonate lid permits viewing of samples without disturbing internal temperature. Opaque lid option available for light sensitive samples.

Shakers can hold up to 4 microplates or deepwell plates or 2 microtube racks.

Certifications: CE marked. RoHS compliant.

Ordering information: Units include a 92” (234cm) detachable, Euro style plug. Includes a NIST traceable calibration certificate by an ISO/IEC 17025 accredited laboratory. Multiple data points within the temperature (incubating model), speed range and time function are reported on the certificate along with the associated uncertainties.



 

ORDER

Expand All / Collapse All
Description Platform Size Overall Dimensions Orbit Speed Temperature Range Supplier No. VWR Catalog Number Unit Price Quantity
Talboys Touch 1000MP Microplate Shaker with NIST Certificate 11 x 7.75" 17 x 11 x 4" 3 mm (1/8") 100–1200 rpm 980TAOMPTSEUC 10822-470 Each Retrieving
Talboys Touch 1000MP Incubating Microplate Shaker with NIST Certificate 11 x 7.75" 17 x 11 x 10.7" 3 mm (1/8") 100–1200 rpm 5° above ambient to 65°C (149°F) 980TAIMPTSEUC 10822-498 Each Retrieving
Talboys Touch 1000MP Incubating Microplate Shaker Opaque Lid with NIST Certificate 11 x 7.75" 17 x 11 x 10.7" 3 mm (1/8") 100–1200 rpm 5° above ambient to 65°C (149°F) 980TAIMOTSEUC 10822-502 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Speed accuracy (%) +/- 2%
Timer 1 min. to 99 hrs., 59 min.
Electrical 230V, 50/60Hz
Max. Load 4 Microplates or 2 Microplate Racks
Platform Material Aluminum

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty