Would you like to log in again?
Rubber Bulb, Saint-Globin
Supplier: Saint Gobain Performance Plastics
DX101012-10 DX101013-10 DX101011-10 DX101010-10
66111-472PK 53.8 USD InStock
66111-472 66111-474 66111-470 66111-468
Rubber Bulb, Saint-Globin
Samplers Gas Samplers
These bulbs have a thickened wall, between which pipettes are placed.
  • They grip the glass firmly and protect it
  • The inner chamber widens out to give an accurate content
  • Sampling can be achived in a minimum amount of time
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty