Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Professional Touch Hotplate Stirrer with NIST Traceable Certificate, 230V, Talboys

Supplier: TALBOYS ENGINEERING/TROEMNER
984TA7CHSTSEUC
  • Intuitive 4.3” color touch screen
  • Programmable for 5 separate, 5 step programs
  • Visibly track program progress through the live status bar
  • On-board help screens
  • Programmable speed and temperature ramping feature (4 options)
  • Includes NIST traceable certificate for temperature, speed, and time
  • 5 year warranty

The Talboys Professional Touch Hotplate-Stirrers are designed for applications that require exceptional accuracy, stability, and repeatability. Exclusive safety features help protect operator and sample. Enhanced microprocessor control offers an external RTD probe option that delivers superior temperature control of the sample. Stirring function, with continuous duty motor and powerful magnet, maintains set speed even under changing load or viscosity. Includes PT1000 RTD temperature probe kit.

Advanced microprocessor regulates the heating and stirring and brings samples to temperature quickly and efficiently. Programmable temperature and speed ramping feature allows user greater control over their assay when changing parameters within a program. Internal RTD accurately measures and controls temperature with minimal overshoot. Precise speed control provides consistent speed control at all speeds. Powerful continuous duty motor delivers dependable and reliable stirring. LCD touch screen enables faster setting of temperature, speed, and time, which can be viewed at once. Display features on-screen help topics. Touch screen is compatible with rubber gloves used in labs. Program control capabilities allow user programmable operation for automated use and memory. Storage for five separate 5-step programs, or unlimited number of programs with the use of the USB. USB port can transfer data to a flash drive for data logging, program storage, and software updates.

Low profile design takes up minimal bench space and fits easily into fume hoods. Rear housing features an off centered, built-in support rod holder with locking knob that accepts the RTD probe kit. Top plate is ceramic and features a 7 x 7” (17.8 x 17.8cm) chemical resistant, reflective surface that is easy to clean.

Hotplate-stirrers are equipped with a variety of innovative safety features designed to facilitate efficient operation and workplace safety. The unit’s housing is constructed of a heat and chemical resistant polymer that remains cool to the touch throughout use. The spill-resistant housing channels fluids away from internal components in the event of a spill. A maximum temperature limiting function ensures the plate temperature will not exceed preset limits, allowing for control of sensitive flash points. A hot symbol warning light is illuminated when the heat is turned on, and remains lit until the top plate is sufficiently cooled. In timed mode, an alarm will sound when time reaches zero, or when the unit reaches set point temperature. Additionally, if the probe disengages from the sample, the heater will automatically shut off. Temperature calibration mode allows user to calibrate unit to an external temperature device for up to 6 separate user defined points. The unit is equipped with a pulse mode feature ideal for quick stirring. Speed ramping feature slowly increases motor speed for improved safety and enhanced coupling. Avoids splashing, improves spin bar control, and provides excellent low end speed control.

Certifications: CE marked. RoHS compliant.

Ordering information: Units include a 92” (234cm) detachable Euro type plug. Supplied with a probe kit which includes an 8” (20.3cm) stainless steel PT1000 RTD temperature probe, 12” (30.5cm) flex arm, interchangeable 3-prong clamp head, 2-prong clamp head, and spring clamp head. A 1.5” (3.8cm) PTFE coated stir bar is included. 5 year limited warranty on parts and labor. NIST traceable calibration certificate by an ISO/IEC 17025 accredited laboratory. Multiple data points within the temperature, speed, and time function are reported on the certificate along with the associated uncertainties.



 

ORDER

Expand All / Collapse All
Electrical Maximum Capacity Speed Range Supplier No. VWR Catalog Number Unit Price Quantity
230V, 1050W, 4.6A 2500 mL 60 to 1600 rpm 984TA7CHSTSEUC 10835-564 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Temperature Stability (%) +/-1%
Speed stability +/-2%
Timer 1 min. to 99 hr. 59 min.
Shipping Weight 5.4 kg (12 lbs.)
Temperature Range Ambient +5 to 500°C
Overall Dimensions 14.8L x 9.9W x 4.3H"
Top plate dimensions 7 x 7" (17.8 x 12.8 cm)
Top plate material Ceramic

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty