Proliant Biologicals’ Bovine Serum Albumin (BSA), Cohn Analog™, Culture Grade is a high purity, lyophilized “Fraction V” powder derived from the plasma of USDA-inspected healthy animals from abattoirs located in the USA. This BSA is isolated from other plasma protein products and lipids by a unique proprietary, non-solvent procedure. The separation process includes a modified Cohn/heat-shock procedure designed to inactivate proteases and other potentially interfering enzymes. Typical purity levels are in excess of 99%. It is high-performance, growth-enhancing preparation of bovine serum albumin for microbial culture and reproductive biology applications.
- High Purity BSA
- Protease Free
- Endotoxin Free
- IgG Free
- Origin Traceability
- Closed Loop Manufacturing
Proliant Biologicals is the only manufacturer of Bovine Serum Albumin who utilizes USDA Edible Grade Plasma and controls the entire process, from collection, through transport and processing.
Application: Growth of fastidious microorganisms such as leptospira and B. bergdorferi. Preserving and extending reproductive cell viability.
Certifications: ISIA Certification, EDQM, & (EC) 1069/2009 and 142/2011 approved by APHIS, Reference Number is IA-TEC-0002
Delivery information: Product should be stored in a cool, dry environment