Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bovine Serum Albumin (BSA), pH 5.2, Standard Grade, Proliant Biologicals, Inc.

Supplier: Proliant Biologicals
68500-10G 68500-25G 68500-500G 68500-100G 68500-250G 68500-1KG 68500-50G
Proliant Biologicals' Albumin, Bovine, pH 5.2, Standard Grade is a high purity, lyophilized “Fraction V” powder derived from the plasma of USDA-inspected healthy animals from abattoirs located in the USA. The albumin is isolated from other plasma protein products and lipids by a unique proprietary, non-solvent procedure. The separation process includes a modified Cohn/heat-shock procedure designed to inactivate proteases and other potentially interfering enzymes. Typical purity levels are in excess of 99%. It is ideal for most diagnostic and research applications.

  • High Purity
  • Protease Free
  • Endotoxin Free
  • IgG Free
  • Origin Traceability
  • Closed Loop Manufacturing

Proliant Biologicals is the only manufactuerer of Bovine Serum Albumin who utilizes USDA Edible Grade Plasma and controls the entire process, from collection, through transport and processing.

Certifications: ISIA Certification, EDQM, & (EC) 1069/2009 and 142/2011 approved by APHIS, Reference Number is IA-TEC-0002




 

ORDER

Expand All / Collapse All
Size Packaging Supplier No. VWR Catalog Number Unit Price Quantity
10 g HDPE Bottle 68500-10G 10842-684 Each (10g) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

25 g HDPE Bottle 68500-25G 10842-778 Each (25g) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

50 g HDPE Bottle 68500-50G 10842-780 Each (50g) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

100 g HDPE Bottle 68500-100G 10842-782 Each (100g) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

250 g HDPE Bottle 68500-250G 10842-784 Each (250g) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

500 g HDPE Bottle 68500-500G 10842-686 Each (500g) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

1 kg HDPE Bottle 68500-1KG 10842-688 Each (1kg) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name Albumin
Protein Synonyms BSAFraction VCohn Fraction VBovine Serum Albumin
Species Bovine
Source Serum
CAS No. 9048-46-8
Protease-free Yes
Endotoxin-free Yes
Grade Standard Grade
Concentration ≥ 96.0%
Formulation White to yellow with tan to green cast lyophilized powder
Storage Conditions Store sealed in a cool, dry environment
Color White to yellow with tan to green cast
pH 5.0–5.6
Form Lyophilized
Physical Form Lyophilized
Application RIA, EIA, Chemiluminescent, Enzyme Stabilizer, etc.

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty