Rt®-Silica BOND Columns (fused silica PLOT), Restek
19784 19786 19785
10849-012EA 889.84 USD
10849-012 10849-010 10849-014
Rt®-Silica BOND Columns (fused silica PLOT), Restek
Restek’s Rt®-Silica BOND columns are robust, versatile, selective PLOT columns that offer excellent performance for the analysis of light hydrocarbons, sulfur gases, and halocarbons above ambient temperature.
  • Versatile column ideal for analysis of light hydrocarbons, sulfur gases, halocarbons, and carbon dioxide.
  • Individually QC tested with sensitive C4 probes to ensure consistent selectivity.
  • Proprietary manufacturing process practically eliminates particle release, reducing downtime due to obstructed FID jets.
  • Bonded silica stationary phase minimizes impact of water, resulting in reproducible retention times for water-containing samples.
  • Stable to 260°C.
In addition, carbon dioxide and other gases can be retained at ambient temperature on this silica-based column
Using Rt®-Silica BOND columns minimizes downtime because Restek’s unique QC testing protocols ensure consistent column-to-column performance. Only Restek measures the selectivity of every column with methyl acetylene and 1,3-butadiene, unsaturated C4 hydrocarbons that are more sensitive selectivity probes than the unsaturated C3 hydrocarbon probes that are used by other manufacturers. When only the C3 hydrocarbons are used for QC testing, columns with large shifts in selectivity for the C4 hydrocarbons can be inadvertently released to market because the C3-only testing procedures do not reveal shifts for the C4 compounds. In addition to more rigorous selectivity testing, Rt®-Silica BOND columns are also tested to confirm efficiency and inertness in order to provide optimal peak shape and response for active analytes.
As with all Restek PLOT columns, Restek's proprietary manufacturing process minimizes particle generation, which reduces the problems commonly associated with released particles, such as signal spikes, valve damage, and clogged FID jets. Even among Restek’s highly robust PLOT columns, this column is exceptionally stable. Also, compared to other PLOT columns, the Rt®-Silica BOND column displays outstanding stability in the presence of water due to its unique bonded silica stationary phase. The combination of rugged manufacturing and rigorous QC testing ensures every Rt®-Silica BOND column will provide optimal performance and reliable results for active unsaturated hydrocarbons.
High loadability, inertness, and consistent selectivity, as well as unmatched robustness at a maximum temperature of 260°C, make the Rt®-Silica BOND column ideal for the analysis of active unsaturated hydrocarbons.