MXT®-S-BOND Columns (Siltek®-treated stainless steel PLOT), Restek
Supplier: Restek
79712-273 79712
10851-362EA 1103.41 USD
10851-362 10851-704
MXT®-S-BOND Columns (Siltek®-treated stainless steel PLOT), Restek
Chromatography Columns
MXT®-S-BOND columns are fabricated from rugged materials to withstand rough handling and deliver robust performance in gas chromatogrpahy processes.
  • Can be made in small coil diameters—perfect for tight spaces
  • Rugged material withstands rough handling and shock
  • Designed for robust performance in GC processes and field instruments
  • Available in 3.5"-coil diameter or 7"-diameter, 11-pin cage
These Siltek®-treated stainless steel columns deliver reliable, consistent performance.