Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rtx®-Biodiesel TG Columns (fused silica), Restek

Supplier: RESTEK CORP
10291 10293 10292 10294
Rtx®-Biodiesel TG columns are fabricated from fused silica to be stable at high temperatures and deliver low column bleed.

  • Linearity for all reference compounds exceeds method requirements
  • Low column bleed at high temperatures.
  • For glycerin and glycerides analysis, according to ASTM D6584 and EN 14105 methods
  • Stable to 350°C

Columns with retention gaps feature Alumaseal® connectors to prevent leaks and extend column life.




 

ORDER

Expand All / Collapse All
Description Column Format Phase I.D. x L Film Thickness Packing Material Supplier No. VWR Catalog Number Unit Price Quantity
Rtx-Biodiesel TG Capillary Biodiesel 0.32 mm x 15 m 0.10 µm fused silica 10293 10803-188 Each Retrieving
Rtx-Biodiesel TG Capillary Biodiesel 0.32 mm x 15 m 0.10 µm fused silica 10294 10059-024 Each Retrieving
Rtx-Biodiesel TG Capillary Biodiesel 0.32 mm x 10 m 0.10 µm fused silica 10291 10849-436 Each Retrieving
Rtx-Biodiesel TG Capillary Biodiesel 0.32 mm x 10 m 0.10 µm fused silica 10292 10059-022 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Description 10803-188 10059-024 10849-436 10059-022
Temperature Limit to 330/380 °C

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty