Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Stx®-CLPesticides Column Kit (0.25mm ID), Restek

Supplier: RESTEK CORP

11190
RK11190EA 1292.69 USD InStock
RK11190
Stx®-CLPesticides Column Kit (0.25mm ID), Restek
Chromatography Columns
Siltek® passivation technology enables these columns to offer unsurpassed inertness and the highest responses for active pesticides.

  • Application-specific columns for organochlorine pesticides and herbicides
  • Baseline separations in less than 10 minutes
  • Siltek® surface deactivation enhances responses for endrin, DDT, methoxychlor
  • Stable to 330°C

Many laboratories analyzing organochlorine pesticides struggle with breakdown and adsorption of endrin, DDT, and methoxychlor caused by active sites throughout the analytical system, problems which are addressed by these columns.

Note: Columns are not preconnected in this kit.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty