Would you like to log in again?
Rt®-CW20M F&F Columns (fused silica), Restek
Supplier: RESTEK CORP
12539 12523
10059-026EA 1034.82 USD InStock
10059-026 10058-380
Rt®-CW20M F&F Columns (fused silica), Restek
Chromatography Columns
Rt®-CW20M F&F columns are designed for use with specific applications of flavor and fragrance compounds.
  • Application-specific columns for flavor and fragrance compounds, specially tested
  • True nonbonded Carbowax® 20M polarity
  • Temperature range: 60°C to 250°C
  • Polar phase; Carbowax® polyethylene glycol—not bonded
These columns are fabricated from durable fused silica.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty