Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rtx®-CLPesticides Column Kit (0.53 mm ID), Restek

Supplier: RESTEK CORP

11197
10850-858EA 1407.42 USD InStock
10850-858
Rtx®-CLPesticides Column Kit (0.53 mm ID), Restek
Chromatography Columns
Improved resolution and faster analyses, compared to 1701 or phenyl phases, make these the pesticide columns of choice.

  • Application-specific columns for organochlorine pesticides and herbicides
  • Low bleed—ideal for high sensitivity GC-ECD or GC-MS analyses
  • Baseline separations in less than 10 minutes.
  • Stable to 320–340°C
  • Analyze EPA method 8081B, 8082A, 8151A, 504.1, 515, 508.1, and 552.2 compounds without time-consuming column changes

Rtx®-CLPesticides columns are specially designed to overcome the coelutions and analyte breakdown typically encountered in chlorinated pesticide analyses for EPA methods 8081, 608, and CLP

Column bleed, measured by ECD, is extremely low at temperatures up to 330°C, which is critical for baking-out the column to remove high-boiling compounds commonly found in pesticide/PCB extracts. An analysis time of less than 10 minutes improves throughput compared to other stationary phases.

Note: Columns are not preconnected in this kit.

By achieving baseline resolution of the 20 target analytes, more accurate qualitative data can be obtained, providing reliable identification without GC-MS.

Caution: Note: Analyzing dirty or derivatized samples can contaminate your column. Restek does not recommend analyzing trace-level pesticide samples following derivatized samples (e.g., Methods 8151A and 552.2) without first performing inlet maintenance. Standard steps include trimming the guard column and changing the inlet liner, O-ring, seal, and septum.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty