Would you like to log in again?
Rtx®-VRX Columns (fused silica), Restek
Supplier: RESTEK CORP
19316 19385 19308 19320 49314 19309 19388 19315 49315 19319
10058-524 10850-678 10058-518 10058-528 10059-568 10058-520 10850-680 10058-522 10059-570 10058-526
Rtx®-VRX Columns (fused silica), Restek
The Rtx®-VRX stationary phase and optimized column dimensions provide low bleed, excellent resolution, and fast analysis times for volatile compounds.
  • Application-specific columns for volatile organic pollutants
  • Excellent for U.S. EPA method 8021 compounds
  • Stable to 260°C
  • Proprietary Crossbond® phase
These columns are constructed from fused silica.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty