Would you like to log in again?
MXT®-65 Columns (Siltek®-treated stainless steel), Restek
Supplier: RESTEK CORP
77023
10851-702EA 709.17 USD InStock
10851-702
MXT®-65 Columns (Siltek®-treated stainless steel), Restek
Chromatography Columns
MXT®-65 columns are good for general-purpose applications involving phenols, triglycerides, and fatty acids.
  • General-purpose columns for phenols, triglycerides, and fatty acids
  • Temperature range: 50°C to 300°C
  • Equivalent to USP G17 phase
  • Mid- to high-polarity phase; Crossbond® diphenyl dimethyl polysiloxane
These columns are constructed of Siltek®-treated stainless steel.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty