Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rtx®-65 Columns (fused silica), Restek

Supplier: RESTEK CORP
17024 17038 17054 17055 17023 17039
The Rtx®-65 phase contains the highest phenyl content of any bonded stationary phase available to improve separation of aromatic compounds through increased phase-analyte interaction.

  • General-purpose columns for phenols, fatty acids, and triglycerides
  • Temperature range: 50°C to 300°C
  • Mid- to high-polarity phase; Crossbond® diphenyl dimethyl polysiloxane

A unique polarity makes these columns ideal for a variety of analyses, from phenols to FAMEs. As a confirmation column for EPA method 604 phenols, an Rtx®-65 column produces a different elution order compared to the primary Rtx®-5 column. Rtx®-65 columns elute FAMEs according to equivalent chain length, similar to bonded Carbowax® columns, but the Rtx®-65 phase does not suffer the thermal stability limitations of other polar stationary phases.




 

ORDER

Expand All / Collapse All
Description Column Format I.D. x L Film Thickness Packing Material Supplier No. VWR Catalog Number Unit Price Quantity
Rtx-65 Capillary 0.32 mm x 30 m 1.00 µm fused silica 17054 RK17054 Each Retrieving
Rtx-65 Capillary 0.25 mm x 30 m 0.50 µm fused silica 17038 RK17038 Each Retrieving
Rtx-65 Capillary 0.25 mm x 30 m 0.25 µm fused silica 17023 RK17023 Each Retrieving
Rtx-65 Capillary 0.53 mm x 30 m 1.00 µm fused silica 17055 RK17055 Each Retrieving
Rtx-65 Capillary 0.32 mm x 30 m 0.25 µm fused silica 17024 RK17024 Each Retrieving
Rtx-65 Capillary 0.32 mm x 30 m 0.50 µm fused silica 17039 RK17039 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Description RK17054 RK17038 RK17023 RK17055 RK17024 RK17039
Temperature Limit 50 to 260/280 °C 50 to 280/300 °C 50 to 300 °C 50 to 250/270 °C 50 to 300 °C 50 to 280/300 °C

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty